Learn More
Cayman Chemical SB-399885 (hydrochloride), Purity: ≥98%, CAS: 402713-81-9, 50 mg
SDP
Catalog No. 502950120
Encompass_Preferred
A 5-HT6 receptor antagonist (Ki = 0.72 nM); >200-fold selective for 5-HT6 receptors over a panel of receptors, ion channels, and enzymes at 10 µM; inhibits 5-HT-induced cAMP accumulation in HeLa cells expressing recombinant human 5-HT6 receptors (pA2 = 7.85); increases the seizure threshold in a rat model of MES-induced seizures; reduces escape latency in the Morris water maze in aged rats and reverses scopolamine-induced impairments in the novel object recognition task in adult rats at 10 mg/kg; has antidepressant and anxiolytic-like effects in mice and rats, respectively
Catalog No. 50-295-0120 Supplier Cayman Chemical Supplier No. 3281750 MG
A 5-HT6 receptor antagonist (Ki = 0.72 nM); >200-fold selective for 5-HT6 receptors over a panel of receptors, ion channels, and enzymes at 10 µM; inhibits 5-HT-induced cAMP accumulation in HeLa cells expressing recombinant human 5-HT6 receptors (pA2 = 7.85); increases the seizure threshold in a rat model of MES-induced seizures; reduces escape latency in the Morris water maze in aged rats and reverses scopolamine-induced impairments in the novel object recognition task in adult rats at 10 mg/kg; has antidepressant and anxiolytic-like effects in mice and rats, respectively