Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-408124 hydrochloride | 1431697-90-3 | 98.8% | 392.83 g/mol | C19H19ClF2N4O | 10 MG
SDP

Catalog No. 5000298473
Encompass_Preferred

SB-408124 hydrochloride is a selective, non-peptide antagonist of the orexin receptor 1 (OX1) used as a research reagent in orexin receptor pharmacology. It binds OX1 with nanomolar affinity and shows approximately 50-fold selectivity over OX2.

  • High purity: 98.8%.
  • Molecular weight: 392.83 g/mol.
  • Chemical formula: C19H19ClF2N4O.
  • Target activity: OX1 receptor antagonist; Ki = 57 nM (whole cell), 27 nM (membrane).
  • Solubility: H2O 1 mg/mL (requires warming/ultrasonic); DMSO <1 mg/mL.
  • Appearance: solid, white to gray.
  • Recommended storage: sealed, away from moisture; in solvent -80°C for long-term storage.

Catalog No. 50-002-98473 Supplier Medchemexpress LLC Supplier No. HY7661210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB-408124 hydrochloride is a selective, non-peptide antagonist of the orexin receptor 1 (OX1) used as a research reagent in orexin receptor pharmacology. It binds OX1 with nanomolar affinity and shows approximately 50-fold selectivity over OX2.

  • High purity: 98.8%.
  • Molecular weight: 392.83 g/mol.
  • Chemical formula: C19H19ClF2N4O.
  • Target activity: OX1 receptor antagonist; Ki = 57 nM (whole cell), 27 nM (membrane).
  • Solubility: H2O 1 mg/mL (requires warming/ultrasonic); DMSO <1 mg/mL.
  • Appearance: solid, white to gray.
  • Recommended storage: sealed, away from moisture; in solvent -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.