Learn More
Medchemexpress LLC SB-408124 Hydrochloride | 1431697-90-3 | 98.8% | C19H19ClF2N4O | 50 MG
SDP
Catalog No. 5000434438
Encompass_Preferred
SB-408124 Hydrochloride is a selective non-peptide orexin receptor 1 (OX1) antagonist, which has demonstrated potent activity with Ki values of 57 nM and 27 nM in whole cell and membrane assays, respectively. It exhibits a high degree of specificity, showing 50-fold selectivity over the OX2 receptor. This compound is intended for research purposes only and is not sold to patients.
  • Selective non-peptide orexin receptor 1 (OX1) antagonist.
  • Ki values of 57 nM (whole cell) and 27 nM (membrane).
  • Exhibits 50-fold selectivity over OX2 receptor.
Catalog No. 50-004-34438 Supplier Medchemexpress LLC Supplier No. HY7661250MG
SB-408124 Hydrochloride is a selective non-peptide orexin receptor 1 (OX1) antagonist, which has demonstrated potent activity with Ki values of 57 nM and 27 nM in whole cell and membrane assays, respectively. It exhibits a high degree of specificity, showing 50-fold selectivity over the OX2 receptor. This compound is intended for research purposes only and is not sold to patients.
  • Selective non-peptide orexin receptor 1 (OX1) antagonist.
  • Ki values of 57 nM (whole cell) and 27 nM (membrane).
  • Exhibits 50-fold selectivity over OX2 receptor.