Learn More
Abcam SB 408124, non-peptide O x 1 antagonist, 10MG
Catalog No. 502706902
Encompass_Preferred
MW 356.4 Da, Purity >97%. Selective, non-peptide orexin receptor 1 (OX1) antagonist (Ki = 26.9 nM). Displays 50-fold selectivity over OX2 (Kb values are 21.7 and 1405 nM for OX1 and OX2, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6902 Supplier Abcam Supplier No. AB14110510MG
MW 356.4 Da, Purity >97%. Selective, non-peptide orexin receptor 1 (OX1) antagonist (Ki = 26.9 nM). Displays 50-fold selectivity over OX2 (Kb values are 21.7 and 1405 nM for OX1 and OX2, respectively).
The product is subject to the following: Abcam Restricted Use Statement