Learn More
Medchemexpress LLC SB-423562 | 351490-27-2 | C26H32N2O4 | 100 MG
SDP
Catalog No. 5000374869
Encompass_Preferred
SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist. It has potential for osteoporosis research.
  • Short-acting calcium-sensing receptor (CaR) antagonist.
  • Potential for osteoporosis research.
  • Related to inflammation or immune system disease.
  • Involved in GPCR/G protein and CaSR signaling pathways.
Catalog No. 50-003-74869 Supplier Medchemexpress LLC Supplier No. HY15105100MG
SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist. It has potential for osteoporosis research.
  • Short-acting calcium-sensing receptor (CaR) antagonist.
  • Potential for osteoporosis research.
  • Related to inflammation or immune system disease.
  • Involved in GPCR/G protein and CaSR signaling pathways.