Learn More
Medchemexpress LLC SB-423562 | 351490-27-2 | C26H32N2O4 | 50 MG
SDP
Catalog No. 5000374877
Encompass_Preferred
SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist with potential for osteoporosis research. It has a molecular weight of 436.54 and appears as a white to off-white solid. It is for research use only.
  • Potential for osteoporosis research
  • Short-acting calcium-sensing receptor (CaR) antagonist
  • White to off-white solid appearance
  • Molecular weight of 436.54
Catalog No. 50-003-74877 Supplier Medchemexpress LLC Supplier No. HY1510550MG
SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist with potential for osteoporosis research. It has a molecular weight of 436.54 and appears as a white to off-white solid. It is for research use only.
  • Potential for osteoporosis research
  • Short-acting calcium-sensing receptor (CaR) antagonist
  • White to off-white solid appearance
  • Molecular weight of 436.54