Learn More
Medchemexpress LLC HY-10431 100mg Medchemexpress, SB-431542 CAS:301836-41-9 Purity:>98%
SDP
Catalog No. 502028750
Encompass_Preferred
Medchemexpress, HY-10431 100mg SB-431542 CAS:301836-41-9 SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8750 Supplier Medchemexpress LLC Supplier No. HY10431100MG
Medchemexpress, HY-10431 100mg SB-431542 CAS:301836-41-9 SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.