Learn More
Apexbio Technology LLC SB 431542, 301836-41-9, MFCD05865244, 10 mg
SDP
Supplier:  Apexbio Technology LLC A824910
Encompass_Preferred
MF: C22H16N4O3, MW: 384.39, Solubility: 19.2mg/mL in DMSO, 10.06 mg/mL in EtOH with ultrasonic,insoluble in H2O. SB431542 is a selective ATP-competitive inhibitor of ALK5 (activin receptor-like kinase 5) with an IC50 value of 94 nM.
Catalog No. 50-101-2514