Learn More
Apexbio Technology LLC SB 431542 301836-41-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A82495.1
Encompass_Preferred
SB 431542 (CAS number 301836-41-9) is a selective ATP-competitive inhibitor targeting activin receptor-like kinase 5 (ALK5) a type I receptor within the transforming growth factor- (TGF- ) signaling pathway It inhibits ALK5 activity with an IC50 value of 94 nM thereby preventing phosphorylation of Smad2 proteins and their subsequent nuclear accumulation Additionally SB 431542 inhibits closely related receptors ALK4 and ALK7 while showing minimal or no significant activity against ALK1 ALK2 ALK3 and ALK6 at relevant concentrations Commonly utilized in cellular assays to investigate TGF- signaling this compound is valuable for studying associated cellular processes and diseases
Catalog No. 50-000-62210