Learn More
Medchemexpress LLC HY-10431 50mg , SB-431542 CAS:301836-41-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1043150MG
Encompass_Preferred
HY-10431 50mg SB-431542 CAS: 301836-41-9 SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9039