Learn More
Medchemexpress LLC SB 452533 | 459429-39-1 | 99.1% | 376.29 | 1 ML
SDP
Catalog No. 5000344278
Encompass_Preferred
SB 452533 is a potent and selective TRPV1 antagonist with a pKb of 7.8. It is for research use only and not sold to patients.
  • Potent antagonist at the cloned recombinant rat TRPV1 receptor in HEK293 cells.
  • Reduces phosphorylation of CAMKK2 and AMPK.
  • Abolishes the effects of small dose Capsaicin in reducing Bleomycin-induced pulmonary fibrosis in mice.
Catalog No. 50-003-44278 Supplier Medchemexpress LLC Supplier No. HY10845850MG
SB 452533 is a potent and selective TRPV1 antagonist with a pKb of 7.8. It is for research use only and not sold to patients.
  • Potent antagonist at the cloned recombinant rat TRPV1 receptor in HEK293 cells.
  • Reduces phosphorylation of CAMKK2 and AMPK.
  • Abolishes the effects of small dose Capsaicin in reducing Bleomycin-induced pulmonary fibrosis in mice.