Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB 452533 | 459429-39-1 | 99.1% | 376.29 g/mol | C18H22BrN3O | 5 MG
SDP

Catalog No. 5000263311
Encompass_Preferred

SB 452533 is a potent, selective TRPV1 antagonist supplied for research use. It is provided with analytical documentation and safety data suitable for pharmacological and preclinical studies of TRPV1-mediated signaling.

  • Selective TRPV1 antagonist for receptor pharmacology studies.
  • Purity 99.11%.
  • Molecular weight 376.29 g/mol.
  • Chemical formula C18H22BrN3O.
  • White to off-white solid physical form.
  • Supplied with COA, LCMS data, and SDS documentation.
  • Soluble in DMSO for assay preparation.

Catalog No. 50-002-63311 Supplier Medchemexpress LLC Supplier No. HY1084585MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB 452533 is a potent, selective TRPV1 antagonist supplied for research use. It is provided with analytical documentation and safety data suitable for pharmacological and preclinical studies of TRPV1-mediated signaling.

  • Selective TRPV1 antagonist for receptor pharmacology studies.
  • Purity 99.11%.
  • Molecular weight 376.29 g/mol.
  • Chemical formula C18H22BrN3O.
  • White to off-white solid physical form.
  • Supplied with COA, LCMS data, and SDS documentation.
  • Soluble in DMSO for assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.