Learn More
TARGETMOL CHEMICALS INC SB-505124 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T246210MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB505124 is a selective inhibitor of TGF(beta)R for ALK4 ALK5. purity: 99%
Catalog No. 50-257-7116