Learn More
Abcam SB 505124, ALK5 inhibitor, 5MG
Catalog No. 502705800
Encompass_Preferred
MW 335.4 Da, Purity >98%. Potent, reversible, competitive ALK5 inhibitor (IC₅₀ values are 47 and 129 nM for ALK5 and ALK4 respectively). Inhibits ALK7 but not ALK1, 2, 3, or 6. Shows antifibrotic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5800 Supplier Abcam Supplier No. AB1444025MG
MW 335.4 Da, Purity >98%. Potent, reversible, competitive ALK5 inhibitor (IC₅₀ values are 47 and 129 nM for ALK5 and ALK4 respectively). Inhibits ALK7 but not ALK1, 2, 3, or 6. Shows antifibrotic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement