Learn More
Apexbio Technology LLC SB-505124 hydrochloride, 10mg. Cas: 356559-13-2 MFCD: MFCD08276927. ALK5/ALK4/ALK7 inhibitor in TGF-β/activin signalling,selective
SDP
Supplier:  Apexbio Technology LLC A379910
Encompass_Preferred
SB505124 is a selective inhibitor of TGFR for ALK4, ALK5 with IC50 value of 129 nM and 47 nM [1].Anaplastic lymphoma kinase (ALK) gets involved in the development of the brain and influence specific neurons in the nervous system. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4841