Learn More
Apexbio Technology LLC SB-505124 hydrochloride 356559-13-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064370
Encompass_Preferred
SB-505124 hydrochloride (CAS 356559-13-2) is a potent selective inhibitor targeting transforming growth factor-beta (TGF ) receptors ALK4 and ALK5 SB-505124 functions by reversibly competing with ATP binding exhibiting IC50 values of 129 nM for ALK4 and 47 nM for ALK5 By blocking these receptor kinases the compound effectively inhibits downstream signaling events such as Smad2/3 phosphorylation and expression of TGF -regulated genes ( -SMA and CTGF) Research demonstrates its applicability in fibrotic disease models including glaucoma filtration surgery by preventing fibroblast activation and extending filtration bleb lifespan in vivo
Catalog No. 50-000-64370 Supplier Apexbio Technology LLC Supplier No. A37995.1
SB-505124 hydrochloride (CAS 356559-13-2) is a potent selective inhibitor targeting transforming growth factor-beta (TGF ) receptors ALK4 and ALK5 SB-505124 functions by reversibly competing with ATP binding exhibiting IC50 values of 129 nM for ALK4 and 47 nM for ALK5 By blocking these receptor kinases the compound effectively inhibits downstream signaling events such as Smad2/3 phosphorylation and expression of TGF -regulated genes ( -SMA and CTGF) Research demonstrates its applicability in fibrotic disease models including glaucoma filtration surgery by preventing fibroblast activation and extending filtration bleb lifespan in vivo