Learn More
Medchemexpress LLC SB-505124 hydrochloride | 356559-13-2 | 99.3% | 371.86 g/mol | C20H22ClN3O2 | 5 MG
SDP
Catalog No. 5000222004
Encompass_Preferred
SB-505124 hydrochloride is the hydrochloride salt of a selective small-molecule inhibitor of TGF-β type I receptors (ALK4, ALK5, ALK7). It is supplied as a solid research reagent with high purity for use in biochemical assays and cellular signaling studies to modulate TGF-β/activin pathways.
  • Selective TGF-β type I receptor inhibitor (ALK4, ALK5, ALK7).
  • High purity suitable for analytical and research use.
  • Hydrochloride salt form, supplied as a solid.
  • Available in small pack sizes for assay development.
  • Reported IC50 values in the low-nanomolar range for ALK4 and ALK5.
Catalog No. 50-002-22004 Supplier Medchemexpress LLC Supplier No. HY13521A5MG
SB-505124 hydrochloride is the hydrochloride salt of a selective small-molecule inhibitor of TGF-β type I receptors (ALK4, ALK5, ALK7). It is supplied as a solid research reagent with high purity for use in biochemical assays and cellular signaling studies to modulate TGF-β/activin pathways.
  • Selective TGF-β type I receptor inhibitor (ALK4, ALK5, ALK7).
  • High purity suitable for analytical and research use.
  • Hydrochloride salt form, supplied as a solid.
  • Available in small pack sizes for assay development.
  • Reported IC50 values in the low-nanomolar range for ALK4 and ALK5.