Learn More
Cayman Chemical SB-505124 (hydrochloride), Purity: ≥98%, CAS: 356559-13-2, 10 mg
SDP
Catalog No. 502945289
Encompass_Preferred
An ALK5 and ALK4 inhibitor (IC50s = 47 and 129 nM, respectively); selective for ALK5 and ALK4 over a panel of 27 kinases at 10 µM; inhibits ALK7-induced increases in phosphorylated SMAD2 levels in COS-1 cells overexpressing ALK7 at 5 µM; inhibits TGF-β-induced but not TNF-α-induced apoptosis in Fao cells at 1 µM; decreases IOP and subconjunctival immune cell infiltration in a rabbit model of trabeculectomy at 5 mg/eye
Catalog No. 50-294-5289 Supplier Cayman Chemical Supplier No. 4205610 MG
An ALK5 and ALK4 inhibitor (IC50s = 47 and 129 nM, respectively); selective for ALK5 and ALK4 over a panel of 27 kinases at 10 µM; inhibits ALK7-induced increases in phosphorylated SMAD2 levels in COS-1 cells overexpressing ALK7 at 5 µM; inhibits TGF-β-induced but not TNF-α-induced apoptosis in Fao cells at 1 µM; decreases IOP and subconjunctival immune cell infiltration in a rabbit model of trabeculectomy at 5 mg/eye