Learn More
BIOGEMS INTERNATIONAL INC SB 525334 10mg
SDP
Catalog No. 501530046
Encompass
SB 525334 is a selective and potent inhibitor of the TGF-Beta receptor 1 (ALK5). It exhibits no inhibition of ALK2, ALK3, and ALK6. It blocks the activation of Smad2/3 by TGF-β1 in renal proximal tubule cells and attenuates bleomycin-induced pulmonary fibrosis.
Catalog No. 50-153-0046 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 356201510MG
SB 525334 is a selective and potent inhibitor of the TGF-Beta receptor 1 (ALK5). It exhibits no inhibition of ALK2, ALK3, and ALK6. It blocks the activation of Smad2/3 by TGF-β1 in renal proximal tubule cells and attenuates bleomycin-induced pulmonary fibrosis.