Learn More
Selleck Chemical LLC SB525334 100mg 356559-20-1
SDP
Catalog No. 508583
Encompass_Preferred
SB525334 is a potent and selective inhibitor of TGF? receptor I (ALK5) with IC50 of 14.3 nM in a cell-free assay, 4-fold less potent to ALK4 than ALK5 and inactive to ALK2, 3, and 6. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-858-3 Supplier Selleck Chemical LLC Supplier No. S1476100MG
SB525334 is a potent and selective inhibitor of TGF? receptor I (ALK5) with IC50 of 14.3 nM in a cell-free assay, 4-fold less potent to ALK4 than ALK5 and inactive to ALK2, 3, and 6. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.