Learn More
Abcam SB-525334, TGFBR1 inhibitor., 25MG
Catalog No. 502702911
Encompass_Preferred
MW 343.4 Da, Purity >98%. Cell permeable, potent, selective transforming growth factor beta 1 receptor (ALK5) (IC₅₀ = 14.3 nM) and ALK4 (IC₅₀ = 58.5 nM) inhibitor, with no activity at ALK2, 3 and 6 (IC₅₀ > 10 μM). Blocks fibrosis and renal injury markers in a puromycin-induced nephritis model. Causes significant attenuation in Smad2/3 nuclear translocation and a decrease in CTGF-expressing cells, myofibroblast proliferation and type 1 collagen deposition resulting in an overall attenuation in bleomycin-induced pulmonary fibrosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-2911 Supplier Abcam Supplier No. AB22204225MG
MW 343.4 Da, Purity >98%. Cell permeable, potent, selective transforming growth factor beta 1 receptor (ALK5) (IC₅₀ = 14.3 nM) and ALK4 (IC₅₀ = 58.5 nM) inhibitor, with no activity at ALK2, 3 and 6 (IC₅₀ > 10 μM). Blocks fibrosis and renal injury markers in a puromycin-induced nephritis model. Causes significant attenuation in Smad2/3 nuclear translocation and a decrease in CTGF-expressing cells, myofibroblast proliferation and type 1 collagen deposition resulting in an overall attenuation in bleomycin-induced pulmonary fibrosis.
The product is subject to the following: Abcam Restricted Use Statement