Learn More
Enzo Life Sciences SB 590885 dihydrochloride (10mg). CAS: 405554-55-4
SDP
Catalog No. 502013392
Encompass_Preferred
Selective for B-Raf against 46 other kinases. SB 590885 dihydrochloride decreases anchorage-independent growth of melanoma cell lines. Inhibits ERK phosphorylation and proliferation in tumor cells expressing B-RafV600E. Alternative name: N,N-dimethyl-2-[4-[(4E)-4-(1-nitroso-2,3-dihydroinden-5-ylidene)-5-(1H-pyridin-4-ylidene)-1H-imidazol-2-yl]phenoxy]ethanamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3392 Supplier Enzo Life Sciences Supplier No. ENZCHM1130010
Selective for B-Raf against 46 other kinases. SB 590885 dihydrochloride decreases anchorage-independent growth of melanoma cell lines. Inhibits ERK phosphorylation and proliferation in tumor cells expressing B-RafV600E. Alternative name: N,N-dimethyl-2-[4-[(4E)-4-(1-nitroso-2,3-dihydroinden-5-ylidene)-5-(1H-pyridin-4-ylidene)-1H-imidazol-2-yl]phenoxy]ethanamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.