Learn More
Medchemexpress LLC Sb-612111 hydrochloride | 371980-98-2 | 99.3% | 454.86 | C24H30Cl3NO | 10 MG
SDP
Catalog No. 5000297972
Encompass_Preferred
SB-612111 hydrochloride is the hydrochloride salt of a selective NOP/ORL-1 receptor antagonist intended for preclinical research. Supplied as a solid with formulation and storage guidance, it is suitable for in vitro pharmacology and in vivo studies when prepared according to published protocols.
  • High purity (99.3%), suitable for research applications.
  • Hydrochloride salt form; chemical formula C24H30Cl3NO; molecular weight 454.86.
  • Readily soluble in DMSO (≈15.62 mg/mL) with established in vivo formulation options.
  • Supplied as a 10 mg solid, white to off-white in appearance.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months).
Catalog No. 50-002-97972 Supplier Medchemexpress LLC Supplier No. HY18618A10MG
SB-612111 hydrochloride is the hydrochloride salt of a selective NOP/ORL-1 receptor antagonist intended for preclinical research. Supplied as a solid with formulation and storage guidance, it is suitable for in vitro pharmacology and in vivo studies when prepared according to published protocols.
  • High purity (99.3%), suitable for research applications.
  • Hydrochloride salt form; chemical formula C24H30Cl3NO; molecular weight 454.86.
  • Readily soluble in DMSO (≈15.62 mg/mL) with established in vivo formulation options.
  • Supplied as a 10 mg solid, white to off-white in appearance.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months).