Learn More
Medchemexpress LLC SB-612111 hydrochloride | 99.6% | 454.86 | 1 ML
SDP
Catalog No. 5000387850
Encompass_Preferred
SB-612111 hydrochloride is a novel and potent opiate receptor-like orphan receptor (ORL-1) antagonist with high affinity for hORL-1 (Ki = 0.33 nM). It demonstrates selectivity for μ-, κ-, and δ-receptors. This compound effectively antagonizes the pronociceptive action of Nociceptin in an acute pain model.
  • Potent ORL-1 antagonist with high affinity.
  • Exhibits selectivity for μ-, κ-, and δ-receptors.
  • Effectively antagonizes pronociceptive action of Nociceptin.
  • Demonstrates antihyperalgesic effects in animal models.
Catalog No. 50-003-87850 Supplier Medchemexpress LLC Supplier No. HY18618A10MM/1ML
SB-612111 hydrochloride is a novel and potent opiate receptor-like orphan receptor (ORL-1) antagonist with high affinity for hORL-1 (Ki = 0.33 nM). It demonstrates selectivity for μ-, κ-, and δ-receptors. This compound effectively antagonizes the pronociceptive action of Nociceptin in an acute pain model.
  • Potent ORL-1 antagonist with high affinity.
  • Exhibits selectivity for μ-, κ-, and δ-receptors.
  • Effectively antagonizes pronociceptive action of Nociceptin.
  • Demonstrates antihyperalgesic effects in animal models.