Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-612111 hydrochloride | 99.6% | 454.86 | 50 MG
SDP

Catalog No. 5000387974
Encompass_Preferred

SB-612111 hydrochloride is a novel and potent opiate receptor-like orphan receptor (ORL-1) antagonist with a high affinity for hORL-1 (Ki=0.33 nM). It exhibits selectivity for μ-, κ- and δ-receptors. It effectively antagonizes the pronociceptive action of Nociceptin in an acute pain model.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (In Vitro): DMSO: 15.62 mg/mL (34.34 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Catalog No. 50-003-87974 Supplier Medchemexpress LLC Supplier No. HY18618A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB-612111 hydrochloride is a novel and potent opiate receptor-like orphan receptor (ORL-1) antagonist with a high affinity for hORL-1 (Ki=0.33 nM). It exhibits selectivity for μ-, κ- and δ-receptors. It effectively antagonizes the pronociceptive action of Nociceptin in an acute pain model.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (In Vitro): DMSO: 15.62 mg/mL (34.34 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.