Learn More
Medchemexpress LLC SB-616234-A | 908601-49-0 | 99.4% | 574.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1947750MG
Encompass_Preferred
SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, which demonstrates anxiolytic and antidepressant activity. It is intended for research use only.
  • Selective 5-HT1B receptor antagonist
  • Orally bioavailable
  • Anxiolytic activity
  • Antidepressant activity
  • High affinity for human 5-HT1B receptors
  • Over 100-fold selective for most molecular targets, except h5-HT1D receptors
  • Acts as a high affinity antagonist in [35S]-GTPγS binding studies
  • Potentiates electrically stimulated [3H]-5-HT release
  • Reverses SKF-99101H-induced hypothermia
  • Produces dose-related anxiolytic effects in maternal separation-induced vocalisation models
  • Causes a dose-dependent inhibition of ex vivo [3H]-GR125743 binding to rat striatal 5-HT1B receptors
Catalog No. 50-003-85951