Learn More
Medchemexpress LLC SB-616234-A | 908601-49-0 | 99.40% | 574.11 | 100 MG
SDP
Catalog No. 5000384955
Encompass_Preferred
SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, demonstrating anxiolytic and antidepressant activity.
  • High affinity for human 5-HT1B receptors.
  • Over 100-fold selectivity for most molecular targets.
  • Acts as a high-affinity antagonist without agonist activity.
  • Produces dose-related anxiolytic effects.
Catalog No. 50-003-84955 Supplier Medchemexpress LLC Supplier No. HY19477100MG
SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, demonstrating anxiolytic and antidepressant activity.
  • High affinity for human 5-HT1B receptors.
  • Over 100-fold selectivity for most molecular targets.
  • Acts as a high-affinity antagonist without agonist activity.
  • Produces dose-related anxiolytic effects.