Learn More
TARGETMOL CHEMICALS INC SB-657510 5MG
SDP
Catalog No. 502581415
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-657510 is a urotensin II (UII) receptor (UT) antagonist. The Ki values of UT for human monkey cat rat and mouse were 61 17 30 65 and 56 nM respectively. SB-657510 plays an anti-inflammatory role by inhibiting UII-induced inflammatory mediators such as adhesion molecules in human vascular endothelial cells and upregulation of cytokines and tissue factors. SB 657510 has a therapeutic effect on diabetes-related atherosclerotic disease in diabetic mouse models. purity: 98%
Catalog No. 50-258-1415 Supplier TARGETMOL CHEMICALS INC Supplier No. T128485MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-657510 is a urotensin II (UII) receptor (UT) antagonist. The Ki values of UT for human monkey cat rat and mouse were 61 17 30 65 and 56 nM respectively. SB-657510 plays an anti-inflammatory role by inhibiting UII-induced inflammatory mediators such as adhesion molecules in human vascular endothelial cells and upregulation of cytokines and tissue factors. SB 657510 has a therapeutic effect on diabetes-related atherosclerotic disease in diabetic mouse models. purity: 98%