Learn More
Abcam SB 699551 dihydrochloride, 5-HT5A receptor antagonist, 50MG
Catalog No. 502707453
Encompass_Preferred
MW 584.66 g/mol, Purity >98%. Selective 5-HT₅A receptor antagonist (pKi values are 8.3, for 5-HT₅A and < 6.0 for 5-HT₁B/D, 5-HT₂A, 5-HT₂C, 5-HT₁A and 5-HT₇ receptors). Increases extracellular 5-HT levels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7453 Supplier Abcam Supplier No. AB12059050MG
MW 584.66 g/mol, Purity >98%. Selective 5-HT₅A receptor antagonist (pKi values are 8.3, for 5-HT₅A and < 6.0 for 5-HT₁B/D, 5-HT₂A, 5-HT₂C, 5-HT₁A and 5-HT₇ receptors). Increases extracellular 5-HT levels.
The product is subject to the following: Abcam Restricted Use Statement