Learn More
Medchemexpress LLC HY-10633 5mg Medchemexpress, SB-705498 CAS:501951-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106335MG
Encompass_Preferred
Medchemexpress, HY-10633 5mg SB-705498 CAS:501951-42-4 SB-705498 is a potent, selective and orally bioavailable transient receptor potential vanilloid 1 (TRPV1) receptor antagonist with a pIC50 of 7.1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8336