Learn More
Medchemexpress LLC SB-743921 hydrochloride | 940929-33-9 | 99.8% | 553.52 | 10 MG
SDP
Catalog No. 5000365550
Encompass_Preferred
SB-743921 hydrochloride is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM.
  • Potently inhibits colony-forming cell (CFC) formation of chronic myeloid leukemia (CML) primary cells.
  • Exhibits slight inhibitory activities on the colony-forming ability of normal bone marrow progenitors.
  • Induces apoptosis of CML primary CD34+ cells, with slight effect on normal CD34+ cells.
  • Displays additive anti-proliferative effect in KCL22 and CML CD34+ cells when combined with imatinib.
  • Overcomes imatinib resistance in CML cells.
  • Inhibits MEK/ERK and AKT signaling in CML cells.
  • Has good oral bioavailability and pharmacokinetics.
  • Induces complete tumor regression in nude mice bearing lung cancer patient xenografts.
Catalog No. 50-003-65550 Supplier Medchemexpress LLC Supplier No. 10MGHY12069
SB-743921 hydrochloride is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM.
  • Potently inhibits colony-forming cell (CFC) formation of chronic myeloid leukemia (CML) primary cells.
  • Exhibits slight inhibitory activities on the colony-forming ability of normal bone marrow progenitors.
  • Induces apoptosis of CML primary CD34+ cells, with slight effect on normal CD34+ cells.
  • Displays additive anti-proliferative effect in KCL22 and CML CD34+ cells when combined with imatinib.
  • Overcomes imatinib resistance in CML cells.
  • Inhibits MEK/ERK and AKT signaling in CML cells.
  • Has good oral bioavailability and pharmacokinetics.
  • Induces complete tumor regression in nude mice bearing lung cancer patient xenografts.