Learn More
Apexbio Technology LLC SB 747651A dihydrochloride 10mg
SDP
Catalog No. 5000090557
Encompass_Preferred
SB 747651A dihydrochloride is a small-molecule inhibitor targeting mitogen- and stress-activated protein kinase 1 (MSK1) It is designed to selectively inhibit MSK1 activity thereby interfering with phosphorylation-mediated signal transduction pathways downstream of MSK1 SB 747651A dihydrochloride exerts its biological activity primarily through competitive inhibition of MSK1 kinase disrupting phosphorylation of the transcription factor CREB and histone H3 Based on these pharmacological properties SB 747651A dihydrochloride holds research potential in inflammation pathways stress-responsive gene regulation cellular proliferation and differentiation and neuronal plasticity
Catalog No. 50-000-90557 Supplier Apexbio Technology LLC Supplier No. B566010
Add to Cart
Add to Cart
SB 747651A dihydrochloride is a small-molecule inhibitor targeting mitogen- and stress-activated protein kinase 1 (MSK1) It is designed to selectively inhibit MSK1 activity thereby interfering with phosphorylation-mediated signal transduction pathways downstream of MSK1 SB 747651A dihydrochloride exerts its biological activity primarily through competitive inhibition of MSK1 kinase disrupting phosphorylation of the transcription factor CREB and histone H3 Based on these pharmacological properties SB 747651A dihydrochloride holds research potential in inflammation pathways stress-responsive gene regulation cellular proliferation and differentiation and neuronal plasticity