Learn More
Medchemexpress LLC SB-747651A dihydrochloride | 1781882-72-1 | 99.1% | 415.32 | 100 MG
SDP
Catalog No. 5000343599
Encompass_Preferred
SB-747651A dihydrochloride is an ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor. It can be used for inflammation research, and also inhibits PRK2, RSK1, p70S6K, and ROCK-II.
  • Inhibits MSK1 with an IC50 of 11 nM.
  • Affects CXCL2-induced intraluminal crawling of neutrophils.
  • Thwarts intraluminal crawling of adherent neutrophils to optimal sites of emigration.
  • Significantly increases transmigration time and detachment time.
  • Inhibits the migration speed of extravascular chemotaxing neutrophils.
Catalog No. 50-003-43599 Supplier Medchemexpress LLC Supplier No. HY11031310MG
SB-747651A dihydrochloride is an ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor. It can be used for inflammation research, and also inhibits PRK2, RSK1, p70S6K, and ROCK-II.
  • Inhibits MSK1 with an IC50 of 11 nM.
  • Affects CXCL2-induced intraluminal crawling of neutrophils.
  • Thwarts intraluminal crawling of adherent neutrophils to optimal sites of emigration.
  • Significantly increases transmigration time and detachment time.
  • Inhibits the migration speed of extravascular chemotaxing neutrophils.