Learn More
Medchemexpress LLC SB-747651A dihydrochloride | 1781882-72-1 | 99.1% | 415.32 | 50 MG
Supplier: Medchemexpress LLC HY110313100MG
SB-747651A dihydrochloride is an ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor. It also inhibits PRK2, RSK1, p70S6K, and ROCK-II. This product is designed for research and analytical applications, particularly in inflammation research.
- Acts as an ATP-competitive MSK1 inhibitor
- Demonstrates an IC50 of 11 nM for MSK1
- Inhibits PRK2, RSK1, p70S6K, and ROCK-II
- Useful for inflammation research