Learn More
Sigma Aldrich Fine Chemicals Biosciences SB-HEK293-Mock-CTRL Negati1EA
Catalog No. 502900309
Encompass
Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. Membrane Preparations for ATPase Assays are suitable for drug-efflux transporter interaction studies based on ATPase activity determination and could be used for differentiation between transporter substrates and inhibitors. Control Membranes with no- or significantly lower transporter activity are available for both Vesicular Transport and ATPase Assays.
Catalog No. 50-290-0309 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SBCT061EA
Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. Membrane Preparations for ATPase Assays are suitable for drug-efflux transporter interaction studies based on ATPase activity determination and could be used for differentiation between transporter substrates and inhibitors. Control Membranes with no- or significantly lower transporter activity are available for both Vesicular Transport and ATPase Assays.