Learn More
Sigma Aldrich Fine Chemicals Biosciences SB MDR1 Hi5 PREDEASY ATPas
Catalog No. 502906069
Encompass
SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVOs PREDEASY(TM) ATPase Assay Kit provides everything necessary to conduct ATPase activation and inhibition assays to better understand the nature of the interaction between a compound and the transporter. By measuring ATPase activity both activation and inhibition of transporters can be investigated.
Catalog No. 50-290-6069 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SBPE013RXN
SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVOs PREDEASY(TM) ATPase Assay Kit provides everything necessary to conduct ATPase activation and inhibition assays to better understand the nature of the interaction between a compound and the transporter. By measuring ATPase activity both activation and inhibition of transporters can be investigated.