Learn More
As One International Inc 10x Standard Buffer, Magnesium free, 10 x 1.5 ml
Catalog No. 501655972
Encompass_Preferred
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.
Catalog No. 50-165-5972 Supplier As One International Inc Supplier No. AO302210
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.