Learn More
As One International Inc 10x Standard Buffer, Magnesium free and Triton free, 10 x 1.5 ml
Supplier:  As One International Inc AO302510
Encompass_Preferred
Standard Buffer is a traditional potassium buffer and has high specificity. However, optimization of primer annealing temperatures and magnesium concentration is required. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.
Catalog No. 50-165-5974