Learn More
Medchemexpress LLC HY-15166 10mg Medchemexpress, SB1317 CAS:937270-47-8 Purity:>98%
SDP
Catalog No. 502029089
Encompass_Preferred
Medchemexpress, HY-15166 10mg SB1317 CAS:937270-47-8 SB1317 is a potent inhibitor of CDK2, JAK2, and FLT3 for the treatment of cancer, with IC50 of 13, 73, and 56 nM for CDK2, JAK2 and FLT3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9089 Supplier Medchemexpress LLC Supplier No. HY1516610MG
Medchemexpress, HY-15166 10mg SB1317 CAS:937270-47-8 SB1317 is a potent inhibitor of CDK2, JAK2, and FLT3 for the treatment of cancer, with IC50 of 13, 73, and 56 nM for CDK2, JAK2 and FLT3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.