Learn More
AdipoGen SB202190,Chemical. CAS: 152121-30-7
SDP
Catalog No. 501146443
Encompass_Preferred
SB202190, AdipoGen Life Sciences. Chemical. CAS: 152121-30-7. Formula: C20H14FN3O. MW: 331.3. Potent and cell permeable p38 MAP kinase inhibitor. Apoptosis inducer. Inhibits p38alpha and beta, but not gamma and delta isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. JNK activator. Autophagic vacuole inducer
Catalog No. 50-114-6443 Supplier AdipoGen Supplier No. AGCR10028M025
SB202190, AdipoGen Life Sciences. Chemical. CAS: 152121-30-7. Formula: C20H14FN3O. MW: 331.3. Potent and cell permeable p38 MAP kinase inhibitor. Apoptosis inducer. Inhibits p38alpha and beta, but not gamma and delta isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. JNK activator. Autophagic vacuole inducer