Learn More
Apexbio Technology LLC SB202190 (FHPI) 152121-30-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063503
Encompass_Preferred
SB202190 (FHPI CAS 152121-30-7) is a selective MAPK inhibitor targeting specifically the p38 isoforms and with reported IC50 values of approximately 50 nM and 100 nM respectively Through inhibition of p38 signaling SB202190 has been observed to stimulate the proliferation of leukemia cell lines including THP-1 and MV4-11 via activation of ERK and C-Raf phosphorylation pathways Additionally SB202190 modulates cellular responses to genotoxic stress such as hydroxyurea-induced premature chromosome condensation and influences histone acetylation status (Lys5 of histone H4) in plant systems Such activities render SB202190 useful for investigating MAP kinase-dependent pathways in cell proliferation and stress responses
Catalog No. 50-000-63503 Supplier Apexbio Technology LLC Supplier No. A16325.1
 Restricted Item
Add to Cart
Add to Cart
SB202190 (FHPI CAS 152121-30-7) is a selective MAPK inhibitor targeting specifically the p38 isoforms and with reported IC50 values of approximately 50 nM and 100 nM respectively Through inhibition of p38 signaling SB202190 has been observed to stimulate the proliferation of leukemia cell lines including THP-1 and MV4-11 via activation of ERK and C-Raf phosphorylation pathways Additionally SB202190 modulates cellular responses to genotoxic stress such as hydroxyurea-induced premature chromosome condensation and influences histone acetylation status (Lys5 of histone H4) in plant systems Such activities render SB202190 useful for investigating MAP kinase-dependent pathways in cell proliferation and stress responses