Learn More
STEMCELL Technologies SB202190, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72632
Encompass
SB202190 is a selective, potent, cell-permeable inhibitor of p38 MAP kinases, inhibiting p38α (SAPK2A, MAPK14) and p38β (SAPK2B, MAPK11) with IC₅₀ values of 50 and 100 nM, respectively (Davies et al.; Jiang et al.). As a pyridinyl imidazole inhibitor, SB202190 directly binds p38 MAP kinases in the ATP binding pocket (Fox et al.).MAINTENANCE AND SELF-RENEWAL· Improves the self-renewal ability of neural stem cells from NPC1-deficient mice (Yang et al.).· Blocks adiponectin-mediated proliferation of hematopoietic stem cells (DiMascio et al.).· Reduces BMP3-mediated proliferation of C3H10T1/2 mesenchymal stem cells (Stewart et al.). DIFFERENTIATION· Induces cardiomyocyte differentiation from human embryonic stem cells (Graichen et al.).
Catalog No. 50-197-5953