Learn More
AdipoGen SB203580,Chemical. CAS: 152121-47-6
SDP
Supplier:  AdipoGen AGCR10030M001
Encompass_Preferred
SB203580, AdipoGen Life Sciences. Chemical. CAS: 152121-47-6. Formula: C21H16FN3OS. MW: 377.4. Cell permeable, specific p38 MAP kinase inhibitor. Binds to the ATP binding site of p38 MAP kinase. T cell proliferation inhibitor. IL-2 production inhibitor. SAPK/JNK inhibitor. COX-1 and -2 inhibitor. Raf-1 activator. Apoptosis enhancer. Antiproliferative. PDK1 inhibitor. Anti-inflammatory. Review. Enhances cardiomyogenesis of human embryonic stem cells (hESCs).
Catalog No. 50-464-591