Learn More
Enzo Life Sciences SB203580, (1mg), CAS Number: 152121-47-6
SDP
Catalog No. NC9041893
Encompass_Preferred
Protein kinase inhibitor. Cell permeable, specific inhibitor of p38 (SAPK2a). Inhibits p38 MAPK stimulation of MAPKAPK2 (IC.The concentrations of SB203580 required to block Akt phosphorylation (IC50= 3-5µM) are only approximately 10-fold higher than those required to inhibit p38 MAP kinase (IC50=0.3-0.5µM). These data define a new activity for this drug and suggest that extreme caution should be used when interpreting data when SB203580 has been used at concentrations above 1-2µM. Non-specific effects include inhibition of COX-1 and -2. Enhances clonal growth of skin epithelial progenitor cells and stimulates neural stem cell proliferation. Alternative Name: 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)1H-imidazole. Formula: C21H16N3FOS. MW: 377.4. CAS Number: 152121-47-6. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: -20°C
Catalog No. NC9041893 Supplier Enzo Life Sciences Supplier No. BMLEI2860001
Protein kinase inhibitor. Cell permeable, specific inhibitor of p38 (SAPK2a). Inhibits p38 MAPK stimulation of MAPKAPK2 (IC.The concentrations of SB203580 required to block Akt phosphorylation (IC50= 3-5µM) are only approximately 10-fold higher than those required to inhibit p38 MAP kinase (IC50=0.3-0.5µM). These data define a new activity for this drug and suggest that extreme caution should be used when interpreting data when SB203580 has been used at concentrations above 1-2µM. Non-specific effects include inhibition of COX-1 and -2. Enhances clonal growth of skin epithelial progenitor cells and stimulates neural stem cell proliferation. Alternative Name: 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)1H-imidazole. Formula: C21H16N3FOS. MW: 377.4. CAS Number: 152121-47-6. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: -20°C