Learn More
AdipoGen SB204990
SDP
Catalog No. 502562207
Encompass_Preferred
Chemical. CAS 154566-12-8. Formula C18H22Cl2O5. MW 389.3. Potent ATP citrate lyase ACLY inhibitor. ATP citrate lyase ACLY, a key enzyme for lipid synthesis, is frequently overexpressed or activated in cancer to promote lipid synthesis and tumor progression. Cell permeable prodrug form of SB201076. Hypolipidemic agent. Fatty acid synthesis and cholesterol synthesis inhibitor in a dose-dependent manner. Useful agent for immunometabolism research. Orally active in vivo. Shown to block fatty acid synthesis and cancer cell growth in pre-clinical studies.
Catalog No. 50-256-2207 Supplier AdipoGen Supplier No. AGCR13697M001
Chemical. CAS 154566-12-8. Formula C18H22Cl2O5. MW 389.3. Potent ATP citrate lyase ACLY inhibitor. ATP citrate lyase ACLY, a key enzyme for lipid synthesis, is frequently overexpressed or activated in cancer to promote lipid synthesis and tumor progression. Cell permeable prodrug form of SB201076. Hypolipidemic agent. Fatty acid synthesis and cholesterol synthesis inhibitor in a dose-dependent manner. Useful agent for immunometabolism research. Orally active in vivo. Shown to block fatty acid synthesis and cancer cell growth in pre-clinical studies.