Learn More
Eurofins Discoverx SB220025, 0.5 mg. (92-1241)
Supplier:  Eurofins Discoverx 921241
Encompass
SB220025 is a synthetic small molecule, and a potent and selective inhibitor of the p38 mitogen-activated protein kinase (MAPK, IC50=60 nM), 50-1000-fold selective over other MAP kinases. Potent inhibitor of angiogenesis, inflammatory cytokine production, and TNF-α production. Provided as 0.5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9762