Learn More
Medchemexpress LLC HY-16711 10mg , SB225002 CAS:182498-32-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1671110MG
Encompass_Preferred
HY-16711 10mg SB225002 CAS: 182498-32-4 SB225002, a potent, selective and non-peptide CXCR2 antagonist, inhibits 125I-IL-8 binding to CXCR2 with an IC50 of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9261