Learn More
Selleck Chemical LLC SB225002 10mg 182498-32-4
SDP
Supplier:  Selleck Chemical LLC S765110MG
Encompass_Preferred
SB225002 is a potent, and selective CXCR2 antagonist with IC50 of 22 nM for inhibiting interleukin IL-8 binding to CXCR2, > 150-fold selectivity over the other 7-TMRs tested. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5075