Learn More
Medchemexpress LLC HY-103154 10mg Medchemexpress, SB228357 CAS:181629-93-6 Purity:>98%
SDP
Catalog No. 501968044
Encompass_Preferred
Medchemexpress, HY-103154 10mg SB228357 CAS:181629-93-6 SB228357 is a selective, potent oral active 5-HT2C/2B receptor antagonist with pKi values of 6.9, 8.0 and 9.0 for 5-HT2A, 5-HT2B and 5-HT2C, respectively. SB228357 has antidepressant/anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8044 Supplier Medchemexpress LLC Supplier No. HY10315410MG
Medchemexpress, HY-103154 10mg SB228357 CAS:181629-93-6 SB228357 is a selective, potent oral active 5-HT2C/2B receptor antagonist with pKi values of 6.9, 8.0 and 9.0 for 5-HT2A, 5-HT2B and 5-HT2C, respectively. SB228357 has antidepressant/anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.