Learn More
Cayman Chemical SB-242084 (hydrochloride), Purity: ≥98%, CAS: 1049747-87-6, 5 mg
SDP
Catalog No. 502890065
Encompass_Preferred
An antagonist of the 5-HT2C receptor (pKi = 9.0), with at least 100-fold more selectivity over other 5-HT, dopamine, or adrenergic receptors; brain penetrant with significant anxiolytic activity; used extensively in animal research
Catalog No. 50-289-0065 Supplier Cayman Chemical Supplier No. 100965 MG
An antagonist of the 5-HT2C receptor (pKi = 9.0), with at least 100-fold more selectivity over other 5-HT, dopamine, or adrenergic receptors; brain penetrant with significant anxiolytic activity; used extensively in animal research